Skip to Content
MilliporeSigma

H8916

Tumor Necrosis Factor-α human

≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

TNF-α

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
human
Recombinant:
expressed in HEK 293 cells
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
17 kDa (glycosylated), ~17.4 kDa
Impurities:
≤1.00 EU/μg (endotoxin)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤1.0 ng/mL ED50

quality

endotoxin tested

mol wt

17 kDa (glycosylated), ~17.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1.00 EU/μg (endotoxin)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TNF(7124)

General description

The TNFα (tumor necrosis factor α) gene is mapped to human chromosome 6p21.33. TNFα is a member of TNF superfamily. TNF-α has a palmitoyl group in the cysteine residue and is phosphorylated in the transmembrane region serine residue.

Application

Tumor Necrosis Factor-a human has been used:
  • as a permeability inducing agent for endothelial cell monolayer permeability assay
  • as a reactive oxygen species inducer in primary rat cardiac microvascular endothelial cells (RCMVECs)
  • in the activation of nuclear factor kappa B (NF-κB) in human embryonic kidney cells (HEK293), neuroblastoma SH-SY5Y cells and HeLa cells
  • in the stimulation of the human keratinocyte cell line(HaCaT) and human coronary artery endothelial cells (HCAECs)
Tumor Necrosis Factor-α (TNF-α) human has been used
  • In induction of netting neutrophils by anti-neutrophil cytoplasmic antibody and to study its effect on platelet activation and formation of monomeric C-reactive protein.
  • To study the effect of TNF-α on miR-221 and fractalkine expression.
  • To induce inflammatory cell responses.
  • In NF-κB luciferase reporter assay.

Biochem/physiol Actions

Tumor Necrosis Factor-α (TNF-α) is a potent pro-inflammatory cytokine that plays a role in the rheumatoid arthritis pathology, psoriatic arthritis (PsA) and psoriasis. It stimulates interleukins IL-1 and IL-6. Polymorphism in the TNF-α gene is associated with destructive arthropathy in PsA. The post-translational modifications in TNF-a is crucial for its functionality.
Tumor necrosis factor-α (TNF-α), also known as cachectin, is expressed as a 26 kDa membrane bound protein and is then cleaved by TNF-α converting enzyme (TACE) to release the soluble 17 kDa monomer, which forms homotrimers in circulation. TNF-α plays roles in anti-tumor activity, immune modulation, inflammation, anorexia, cachexia, septic shock, viral replication and hematopoiesis. TNF-α is cytotoxic for many transformed cells, but in normal diploid cells, it stimulates proliferation (fibroblasts), differentiation (myeloid cells) or activation (neutrophils). TNF-α also shows antiviral effects against both DNA and RNA viruses and induces production of several other cytokines.
Tumore necrosis factor-α (TNF-α), also known as cachectin, is expressed as a 26 kDa membrane bound protein and is then cleaved by TNF-α converting enzyme (TACE) to release the soluble 17 kDa monomer, which forms homotrimers in circulation. TNF-α plays roles in antitumor activity, immune modulation, inflammation, anorexia, cachexia, septic shock, viral replication and hematopoiesis. TNF-α is expressed by a great variety of cells, with numerous inductive and suppressive agents. Primarily, TNF-α is produced by macrophages in response to immunological challenges such as bacteria (lipopolysaccharides), viruses, parasites, mitogens and other cytokines. TNF-α is cytotoxic for many transformed cells (its namesake activity) but in normal diploid cells, it can stimulate proliferation (fibroblasts), differentiation (myeloid cells) or activation (neutrophils). TNF-α also shows antiviral effects against both DNA and RNA viruses and it induces production of several other cytokines. Although TNF-α is used in clinical trials as an antitumor agent, Sigma′s cytokine, growth factor and hormone products are for research only. TNF-α and the related molecule TNF-β (LT-α) share close structural homology with 28% amino acid sequence identity and both activate the same TNF receptors, TNFR1 and TNFR2. Mouse and human TNF-α share 79% amino acid sequence identity. Unlike human TNF-α, the mouse form is N-glycosylated.

Preparation Note

Tumor Necrosis Factor-α (TNF-α) is expressed in human HEK 293 cells and has been shown to be predominantly a glycosylated, non-covalently linked homotrimer with a molecular mass of 51 kDa (gel filtration). Production in human 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent cytotoxity of the TNF alpha sensitive cell line L-929 in the presence of actinomycin D.


Still not finding the right product?

Explore all of our products under Tumor Necrosis Factor-α human


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library