Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3839

Sigma-Aldrich

Danirixin

≥98% (HPLC)

Synonym(s):

3-{4-Chloro-2-hydroxy-3-[(3S)-piperidine-3-sulfonyl]phenyl}-1-(3-fluoro-2-methylphenyl)urea, GSK 1325756, GSK 1325756B, GSK-1325756, GSK-1325756B, GSK1325756, GSK1325756B, N-[4-Chloro-2-hydroxy-3-[(3S)-3-piperidinylsulfonyl]phenyl]-N′-(3-fluoro-2-methylphenyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClFN3O4S
CAS Number:
Molecular Weight:
441.90
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C(NC1=CC=C(C(S([C@H]2CCCNC2)(=O)=O)=C1O)Cl)NC3=C(C(F)=CC=C3)C

Biochem/physiol Actions

Danirixin (GSK1325756) is an orally active, selective CXC chemokine receptor 2 (CXCR2) antagonist with 78-fold lower affinity toward CXCR1 (CXCR2/CXCR1 IC50 = 12.5/977 nM by competitive binding against 0.225 nM [125I] CXCL8). Danirixin blocks agonist-induced neutrophil CD11b expression in vitro (IC50 = 420/890 nM against induction by 10 nM human CXCL1/rat CXCL2 using rat/human whole blood) and prevents lung neutrophils influx in rats in vivo following aerosol lipopolysaccharide or ozone challenge (ED50 = 1.4 and 16 mg/kg, respectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Nie et al.
International immunopharmacology, 95, 107153-107153 (2021-03-08)
Breast cancer is the most frequent cancer among females and the second most common cause of cancer deaths worldwide. Tumor-associated macrophages (TAMs) are the most abundant immune cell population in the tumor microenvironment, including breast cancer. Breast cancer stem cells
Danirixin: A Reversible and Selective Antagonist of the CXC Chemokine Receptor 2
Journal of Pharmacology and Experimental Therapeutics, 362(2), 338-346 (2017)
CXCL-8-dependent and -independent neutrophil activation in COPD: experiences from a pilot study of the CXCR2 antagonist danirixin.
ERJ open research, 6(4), 00583-2020 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service