Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O2260

Sigma-Aldrich

Mouse Osteopontin

recombinant, expressed in NSO cells, powder, suitable for cell culture

Synonym(s):

Bone Sialoprotein-1, Early T Lymphocyte Activation Protein-1 (ETA-1), OPN, Secreted Phosphoprotein 1 (Spp1)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Osteopontin from mouse, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

sterility

sterile

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

30 kDa by SDS-PAGE (reducing)
65 kDa by SDS-PAGE (reducing) (doublet)

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Spp1(20750)

Application

Osteopontin from mouse has been used:
  • to study its role in a mouse model of preterm brain injury
  • as a positive control to analyze the relationship between the vascular calcification process and osteopontin (OPN) overexpression in the neointima of atherosclerotic plaques
  • to study its interaction on the crystal growth process of hydroxyapatite in agarose hydrogels

Biochem/physiol Actions

Osteopontin (OPN) is a secreted, highly acidic, calcium binding, phosphorylated glycoprotein that is chemotactic for macrophages, smooth muscle cells, endothelial cells and glial cells. In activated macrophages, OPN inhibits nitric oxide production and cytotoxicity. Human, mouse, rat, pig and bovine OPN share 40% amino acid sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to mediate 293 cell adhesion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osteopontin as a means to cope with environmental insults: regulation of inflammation, tissue remodeling, and cell survival
Denhardt DT, et al.
The Journal of Clinical Investigation, 107(9), 1055-1061 (2001)
The effect of osteopontin and osteopontin-derived peptides on preterm brain injury
Albertsson AM, et al.
Journal of Neuroinflammation, 11(1), 197-197 (2014)
Osteopontin upregulation in atherogenesis is associated with cellular oxidative stress triggered by the activation of scavenger receptors
Jimenez CAE, et al.
Archives of Medical Research, 43(2), 102-111 (2012)
Effect of osteopontin, chondroitin sulfates (B, C), and human serum albumin on the crystallization behavior of hydroxyapatite in agarose and silica hydrogels
Jimenez CAE, et al.
Crystal Growth & Design, 8(4), 1335-1339 (2008)
Suhail Tahir et al.
Journal of immunology (Baltimore, Md. : 1950), 194(12), 5725-5735 (2015-05-15)
Immune aging results in diminished adaptive immunity and increased risk for autoimmunity. We previously reported a unique PD-1(+) CD44(high)CD4(+) T cell population that increases with age in normal mice. In this study, we indicate that the age-dependent PD-1(+) CD44(high)CD4(+) T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service