Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0289

Sigma-Aldrich

PF-06651481-00

≥98% (HPLC)

Synonym(s):

4-[(3,5-Dichloro-4-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile, Bosutinib isomer 4, PF-06651481

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29Cl2N5O3
CAS Number:
Molecular Weight:
530.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CN(CC1)CCN1CCCOC2=C(OC)C=C3C(N=CC(C#N)=C3NC4=CC(Cl)=C(OC)C(Cl)=C4)=C2

InChI

1S/C26H29Cl2N5O3/c1-32-6-8-33(9-7-32)5-4-10-36-24-14-22-19(13-23(24)34-2)25(17(15-29)16-30-22)31-18-11-20(27)26(35-3)21(28)12-18/h11-14,16H,4-10H2,1-3H3,(H,30,31)

InChI key

YCLIWTLPTXAGPQ-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06651481-00 is an isomer of orally available dual Src/Abl kinase inhibitor Bosutinib.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service