Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3294

Sigma-Aldrich

SGC-CK2-1N

≥98% (HPLC)

Synonym(s):

N-(5-((3-Cyano-7-(cyclopropyl(methyl)amino)pyrazolo[1,5-a]pyrimidin-5-yl)(methyl)amino)-2-methylphenyl)cyclopropanecarboxamide, SGC-CK2-1 inactive control, SGC-CK2-1 negative control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N7O
CAS Number:
Molecular Weight:
415.49
UNSPSC Code:
12352200
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

SGC-CK2-1N is an inactive structure analog of and the recommended negative control for the potent and highly selective casein 2 (CK2, CKII) inhibitor SGC-CK2-1 (CK2α/CK2α’ IC50 >10 μM with SGC-CK2-1N vs IC50 = 4.2/2.3 nM with SGC-CK2-1 by cell-free enzyme assay).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service