Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H9NO2S
Molecular Weight:
207.25
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
liquid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Quality Level
assay
≥95% (HPLC)
form
liquid
potency
10 μM IC50
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
shipped in
wet ice
storage temp.
2-8°C
General description
Potential anti-inflammatory agent. Selectively and irreversibly inhibits the TNFα-inducible phosphorylation of IκB-α (IC50 = 10 µM) without affecting the constitutive IκB-α phosphorylation. Decreases nuclear translocation of NFκB and inhibits TNFα-induced surface expression of the endothelial-leukocyte cell adhesion molecules E-selectin, VCAM-1, and ICAM-1. E-selectin inhibition was shown to be irreversible (IC50 ~10 µM). Also reversibly increased the activity of p38 kinase and JNK-1 and stimulated tyrosine phosphorylation of a 130-140 kDa protein.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
TNF-α-inducible phosphorylation of IκBα
TNF-α-inducible phosphorylation of IκBα
Product does not compete with ATP.
Reversible: no
Packaging
Packaged under inert gas
Physical form
Supplied as a 100 mM (10 mg/483 µl) solution of BAY 11-7082 (Cat. No. 196870) in DMSO.
Other Notes
Kamthong PJ. and Wu, M-C. 2001. Biochem. J. 356.
Izban, K.F., et al. 2000. Hum. Pathol.31, 1482.
Pierce, J.W., et al. 1997. J. Biol. Chem. 272, 21096.
Izban, K.F., et al. 2000. Hum. Pathol.31, 1482.
Pierce, J.W., et al. 1997. J. Biol. Chem. 272, 21096.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Irritant (B)
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
188.6 °F - closed cup - (Dimethylsulfoxide)
flash_point_c
87 °C - closed cup - (Dimethylsulfoxide)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Miriam Paz-Barba et al.
Diabetologia, 67(1), 124-136 (2023-11-04)
Inflammation induces beta cell dysfunction and demise but underlying molecular mechanisms remain unclear. The apolipoprotein L (APOL) family of genes has been associated with innate immunity and apoptosis in non-pancreatic cell types, but also with metabolic syndrome and type 2
Mariana Sponchiado et al.
Physiological reports, 9(3), e14749-e14749 (2021-02-14)
Substance P (SP) is a tachykinin that regulates airway mucous secretion in both health and disease. Our study aimed to determine whether overexpression of SP without pre-existing inflammation was sufficient to induce changes in mucin secretion and transport in small
Charlotte Kötting et al.
Cancers, 13(6) (2021-04-04)
Curcumin is known to have immune-modulatory and antitumor effects by interacting with more than 30 different proteins. An important feature of curcumin is the inhibition of nuclear factor kappa of activated B-cells (NF-κB). Here, we evaluate the potential of curcumin
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 196871-10MG | 04055977206579 |