Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

662009

Sigma-Aldrich

U0126

InSolution, ≥98%, A potent and specific inhibitor of MEK1 and MEK2

Synonym(s):

InSolution U0126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N6S2 · 0.5C2H6O
CAS Number:
Molecular Weight:
403.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−70°C

SMILES string

S(c2c(cccc2)N)\C(=C(\C(=C(\Sc1c(cccc1)N)/N)\C#N)/C#N)\N

InChI

1S/C18H16N6S2/c19-9-11(17(23)25-15-7-3-1-5-13(15)21)12(10-20)18(24)26-16-8-4-2-6-14(16)22/h1-8H,21-24H2/b17-11+,18-12+

InChI key

DVEXZJFMOKTQEZ-JYFOCSDGSA-N

General description

A potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK. Has very little effect on other kinases such as Abl, Cdk2, Cdk4, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, PKC, and Raf. Also acts as an immunosuppressant by effectively blocking IL-2 synthesis and T cell proliferation without affecting the long-term outcomes of either T cell activation or tolerance. Reported to undergo rapid isomerization in DMSO and exist as an equilibrium mixture of (Z,Z), E,E) and (Z,E) isomers with minimal loss of activity over a period of 1 month.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 100 mM (5 mg/124 µl) solution of U0126 (Cat. No. 662005) in DMSO.

Reconstitution

Following initial thaw, aliquot into precooled vials, freeze immediately at (-70°C).

Other Notes

Duncia, J.V., et al. 1998. Bioorg. Med. Chem. Lett.8, 2839.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamish T J Gilbert et al.
JOR spine, 7(1), e1315-e1315 (2024-01-22)
Stem cell-based therapies show promise as a means of repairing the degenerate intervertebral disc, with growth factors often used alongside cells to help direct differentiation toward a nucleus pulposus (NP)-like phenotype. We previously demonstrated adipose-derived stem cell (ASC) differentiation with
Ting-Ting Wang et al.
Experimental dermatology, 33(7), e15128-e15128 (2024-07-08)
Dry skin is common to many pruritic diseases and is difficult to improve with oral traditional antihistamines. Recently, increasing evidence indicated that histamine H4 receptor (H4R) plays an important role in the occurrence and development of pruritus. Extracellular signal-regulated kinase
Yiming Zhang et al.
International journal of molecular medicine, 46(3), 965-976 (2020-06-26)
MicroRNAs (miRNAs) are known to have regulatory roles in the osteogenic differentiation of various mesenchymal stem cells (MSCs), although their regulatory role on human adipose‑derived mesenchymal stem cells (hADSCs) remains unclear. The aim of the present study was to investigate
Andrew D Esteves et al.
Journal of virology, 96(5), e0175221-e0175221 (2022-01-06)
Infection of peripheral axons by alpha herpesviruses (AHVs) is a critical stage in establishing a lifelong infection in the host. Upon entering the cytoplasm of axons, AHV nucleocapsids and associated inner-tegument proteins must engage the cellular retrograde transport machinery to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service