Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1896

Sigma-Aldrich

VH298

≥98% (HPLC)

Synonym(s):

(2S,4R)-1-((S)-2-(1-Cyanocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33N5O4S
CAS Number:
Molecular Weight:
523.65
UNSPSC Code:
12352200
NACRES:
NA.77

ligand

VH298

Quality Level

Assay

≥98% (HPLC)

form

powder

reaction suitability

reagent type: ligand

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)C4(CC4)C#N)C(C)(C)C)C

InChI key

NDVQUNZCNAMROD-RZUBCFFCSA-N

Biochem/physiol Actions

VH298 is a cell-permeable small molecule that disrupts the interaction of VHL with Hypoxia-inducible factor HIF-α, stabilizing HIF-α and inducing a hypoxic response.
VH298 is a cell-permeable small molecule that disrupts the interaction of VHL with Hypoxia-inducible factor HIF-α, stabilizing HIF-α and inducing a hypoxic response. Hypoxia-inducible factors (HIFs) are oxygen-sensitive transcription factors that regulate hypoxic signalling. They are regulated by oxygen-dependent prolyl hydroxylase domain (PHD) enzymes and polyubiquitination by the von Hippel–Lindau (VHL) Cullin RING E3 ubiquitin ligase complex. Unlike many PHD inhibitors, which have off-target effects, VH298 acts downstream of the PHD enzymes, inhibiting the VHL:HIF-α interaction. VH298 had a Kd of 90 nM. VH298 was shown to induce HIF transcriptional activity, to increase accumulation of HIF-α in HeLa cells and to stimulate erythropoietin production in RCC4 cells.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julianty Frost et al.
Nature communications, 7, 13312-13312 (2016-11-05)
Chemical strategies to using small molecules to stimulate hypoxia inducible factors (HIFs) activity and trigger a hypoxic response under normoxic conditions, such as iron chelators and inhibitors of prolyl hydroxylase domain (PHD) enzymes, have broad-spectrum activities and off-target effects. Here
Olena S Tokareva et al.
Nature communications, 14(1), 6992-6992 (2023-11-02)
Molecules that induce novel interactions between proteins hold great promise for the study of biological systems and the development of therapeutics, but their discovery has been limited by the complexities of rationally designing interactions between three components, and because known
Yong Hwan Kim et al.
Molecules (Basel, Switzerland), 29(2) (2024-01-23)
Autophagy is a pivotal biological process responsible for maintaining the homeostasis of intracellular organelles. Yet the molecular intricacies of peroxisomal autophagy (pexophagy) remain largely elusive. From a ubiquitin-related chemical library for screening, we identified several inhibitors of the Von Hippel-Lindau
Lauren M Meyers et al.
Toxicology and applied pharmacology, 445, 116041-116041 (2022-05-04)
Transcription factors HIF1 and HIF2 are central regulators of physiological responses to hypoxia and important for normal functioning of tissue stem cells and maintenance of healthy microvasculature. Even modest decreases in HIF activity exert detrimental effects in tissues although it
Misty Shuo Zhang et al.
Nature communications, 13(1), 954-954 (2022-02-19)
Hepatocellular carcinoma (HCC) invariably exhibits inadequate O2 (hypoxia) and nutrient supply. Hypoxia-inducible factor (HIF) mediates cascades of molecular events that enable cancer cells to adapt and propagate. Macropinocytosis is an endocytic process initiated by membrane ruffling, causing the engulfment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service