Skip to Content
MilliporeSigma
All Photos(2)

Documents

904724

Sigma-Aldrich

Pomalidomide-PEG2-butyl iodide

≥95%

Synonym(s):

Crosslinker–E3 Ligase ligand conjugate, Pomalidomide-2-2-6-I, N-(2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)-2-(2-((6-iodohexyl)oxy)ethoxy)acetamide, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28IN3O7
CAS Number:
Molecular Weight:
585.39
UNSPSC Code:
12352101
NACRES:
NA.22

ligand

pomalidomide

Assay

≥95%

form

powder or crystals

reaction suitability

reactivity: sulfuryl reactive
reagent type: ligand-linker conjugate

functional group

alkyl halide

storage temp.

2-8°C

SMILES string

ICCCCCCOCCOCC(NC1=CC=CC2=C1C(N(C3CCC(NC3=O)=O)C2=O)=O)=O

Application

Protein degrader builiding block Pomalidomide-PEG2-butyl Iodide enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a Cereblon (CRBN)-recruiting ligand, a linker with both hydrophobic and hydrophilic moieties, and a pendant iodoalkane for reactivity with a nucleophilic group on a target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant iodo group, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service