Skip to Content
MilliporeSigma
All Photos(1)

Documents

400091

Sigma-Aldrich

DMOG

≥98% (HPLC), solid, HIF-hydroxylase inhibitor, Calbiochem

Synonym(s):

HIF-Hydroxylase Inhibitor, DMOG, N-(Methoxyoxoacetyl)-glycine methyl ester, Dimethyloxalylglycine, HIF Prolyl Hydroxylase Inhibitor I, HIF Aspartyl β-Hydroxylase Inhibitor, HIF Asparanginyl β-Hydroxylase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO5
CAS Number:
Molecular Weight:
175.14
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

HIF-Hydroxylase Inhibitor, DMOG, The HIF-Hydroxylase Inhibitor, DMOG, also referenced under CAS 89464-63-1, controls the biological activity of HIF-Hydroxylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Quality Level

Assay

≥98% (HPLC)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

pink-white to peach-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C6H9NO5/c1-11-4(8)3-7-5(9)6(10)12-2/h3H2,1-2H3,(H,7,9)

InChI key

BNJOZDZCRHCODO-UHFFFAOYSA-N

General description

A cell-permeable 2-OG (2-oxoglutarate) analog that is expected to act as a competitive inhibitor against all 2-OG-dependent dioxygenases. Effectively upregulates normoxia HIF-α transcription activity (effective conc. = 1 mM in cultures) by inhibiting HIF-PH- (HIF-α-prolyl hydroxylase) catalyzed ODD (oxygen-dependent degradation domain) Prolyl hydroxylation (Pro564 and Pro530 in HIF-1α and HIF-2α, respectively) and aspartyl/asparanginyl β-hydroxylase-catalyzed CAD Asp hydroxylation (Asn803 and Asn851 in HIF-1α and HIF-2α, respectively), resulting in a blockage of pVHL (von Hippel-Lindau) ubiquitin ligase complex-mediated HIF-α degradation and a simultaneous promotion of HIF CAD (COOH-terminal activation domain) p300/CBP association. DMOG is also demonstrated to augment artery ligation-induced up-regulation of HIF-1α and FLK (by 100% and 40%, respectively, in 11 days; 8 mg/0.5 ml/animal/i.p.; q.o.d.) in muscle tissues in a murine ischaemic model in vivo.
A cell-permeable 2-OG (2-oxoglutarate) analog that is expected to act as a competitive inhibitor against all 2-OG-dependent dioxygenases. Effectively upregulates normoxia HIF-α transcription activity (effective conc. = 1 mM in cultures) by inhibiting HIF-PH- (HIF-α-prolyl hydroxylase) catalyzed ODD (oxygen-dependent degradation domain) Prolyl hydroxylation (Pro564 and Pro530 in HIF-1α and HIF-2α, respectively) and aspartyl/asparanginyl β-hydroxylase-catalyzed CAD Asp hydroxylation (Asn803 and Asn851 in HIF-1α and HIF-2α, respectively), resulting in a blockage of pVHL (von Hippel-Lindau) ubiquitin ligase complex-mediated HIF-α degradation and a simultaneous promotion of HIF CAD (COOH-terminal activation domain) p300/CBP association. DMOG is also demonstrated to augment artery ligation-induced up-regulation of HIF-1α and FLK (by 100% and 40%, respectively, in 11 days; 8 mg/0.5 ml/animal/i.p.; q.o.d.) in muscle tissues in a murine ischaemic model in vivo.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pollard, P.J., et al. 2008. Biochem. J.416, 387.
Chen, H., et al. 2006. Cancer Res.66, 9009.
Milkiewicz, M., et al. 2004. J. Physiol.560, 21.
Lando, D., et al. 2002. Science295, 858.
Jaakkola, P., et al. 2001. Science292, 468.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Gu et al.
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 98(7), 361-377 (2022-08-01)
Previous studies showed that lanthanum hydroxide (LH) has a therapeutic effect on chronic kidney disease (CKD) and vascular calcification, which suggests that it might have clinical value. However, the target and mechanism of action of LH are unclear. Metabolomics of
Yixi He et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 43(2_suppl), 78-94 (2023-05-13)
Vascular cognitive impairment and dementia (VCID) is a series of cognitive dysfunction associated with cerebrovascular diseases and currently lacks effective treatments. The white matter, which is essential for neuronal information processing and integration, is nourished by a network of capillaries
David Danielpour et al.
Current research in pharmacology and drug discovery, 3, 100076-100076 (2022-01-11)
The imidazolium compound Sepantronium Bromide (YM155) successfully promotes tumor regression in various pre-clinical models but has shown modest responses in human clinical trials. We provide evidence to support that the hypoxic milieu of tumors may limit the clinical usefulness of
Shengnan Wang et al.
Cell biology international, 46(8), 1275-1287 (2022-05-12)
The main reason for the high incidence of cardiovascular disease in chronic kidney disease (CKD) patients with vascular calcification (VC) is also the main cause of death in CKD patients. Lanthanum hydroxide (LH) has an inhibitory effect on VC in
Frederike Boos et al.
Basic research in cardiology, 118(1), 5-5 (2023-01-27)
Long non-coding RNAs (lncRNAs) can act as regulatory RNAs which, by altering the expression of target genes, impact on the cellular phenotype and cardiovascular disease development. Endothelial lncRNAs and their vascular functions are largely undefined. Deep RNA-Seq and FANTOM5 CAGE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service