Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

276863

Sigma-Aldrich

Diethylaluminum cyanide solution

1.0 M in toluene

Synonym(s):

Cyanodiethylalane, Cyanodiethylaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2AlCN
CAS Number:
Molecular Weight:
111.12
Beilstein:
3587410
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in toluene

density

0.864 g/mL at 25 °C

SMILES string

CC[Al](CC)C#N

InChI

1S/2C2H5.CN.Al/c3*1-2;/h2*1H2,2H3;;

InChI key

KWMUAEYVIFJZEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used to hydrocyanate side chain ketones directed by aryl sulfinyl groups.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron, 62, 1245-1245 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service