Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1405

Sigma-Aldrich

MS37452

≥98% (HPLC)

Synonym(s):

1-(2,3-Dimethoxybenzoyl)-4-[(3-methylphenoxy)acetyl]-piperazine (9CI), 1-[4-(2,3-Dimethoxybenzoyl)-1-piperazinyl]-2-(3-methylphenoxy)-ethanonemethylphenoxy)acetyl]-piperazine (9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O5
CAS Number:
Molecular Weight:
398.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(COC1=CC(C)=CC=C1)N2CCN(C(C3=CC=CC(OC)=C3OC)=O)CC2

InChI

1S/C22H26N2O5/c1-16-6-4-7-17(14-16)29-15-20(25)23-10-12-24(13-11-23)22(26)18-8-5-9-19(27-2)21(18)28-3/h4-9,14H,10-13,15H2,1-3H3

InChI key

LUMCNRKHZRYQOV-UHFFFAOYSA-N

Biochem/physiol Actions

MS37452 is a selective Chromobox homolog 7 (CBX7) modulator that disrupts CBX7ChD binding to H3K27me3. MS37452 inhibits CBX7 binding to INK4/ARF gene locus, and induces decline of p14/ARF and p16/INK4a in human PC3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Ren et al.
Chemistry & biology, 22(2), 161-168 (2015-02-11)
Chromobox homolog 7 (CBX7) plays an important role in gene transcription in a wide array of cellular processes, ranging from stem cell self-renewal and differentiation to tumor progression. CBX7 functions through its N-terminal chromodomain (ChD), which recognizes trimethylated lysine 27

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service