Skip to Content
MilliporeSigma

SML0559

LDN193189 hydrochloride

≥98% (HPLC), powder, ALK2/3 inhibitor

Synonym(s):

4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]-quinoline hydrochloride, DM-3189

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H22N6 · xHCl
CAS Number:
Molecular Weight:
406.48 (free base basis)
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

LDN193189 hydrochloride, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to orange

solubility

H2O: 5 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

C(C=N1)(C2=CC=C(N3CCNCC3)C=C2)=CN4C1=C(C5=CC=NC6=C5C=CC=C6)C=N4.Cl

InChI

1S/C25H22N6.ClH/c1-2-4-24-22(3-1)21(9-10-27-24)23-16-29-31-17-19(15-28-25(23)31)18-5-7-20(8-6-18)30-13-11-26-12-14-30;/h1-10,15-17,26H,11-14H2;1H

InChI key

PCCDKTWDGDFRME-UHFFFAOYSA-N

Application

LDN193189 hydrochloride has been used as an ALK2/3 (type I receptor serine-threonine kinases) inhibitor, to study the effect of TGFβ1/2/3 (tumor growth factor β) and BMP (bone morphogenetic protein) signaling on spinal cord development in zebrafish. It has also been used to inhibit SMAD (homologues of the Drosophila protein, mothers against decapentaplegic), in order to prevent non neuronal differentiation.

Biochem/physiol Actions

LDN193189 is a derivative of dorsomorphin that is a highly selective antagonist of BMP receptor isotypes ALK2 and ALK3 (IC50 of: 5 and 30 nM). The selectivity of LDN193189 for ALK2/3 is 200 fold over the TGF-B type receptors ALK4,-5 and -7. In murine smooth muscle cells, the compound inhibits BMP-4 induced phosphorylation of SMAD 1/5/8 with an IC50 of 5 nM.
LDN193189 is a selective BMP receptor antagonist.
Although LDN193189 is a structural analog of dorsomorphin, these two drugs are found to establish different cellular responses. In vitro analysis reveals that LDN193189 inhibits a number of intracellular kinases such as, mitogen activated protein kinase 14 and 8 ( p38and c-Jun N-terminal kinase respectively), as well as those associated with AKT (serine/threonine kinase) and mTOR (mammalian target of rapamycin) signaling mechanisms. LDN193189 is known to elevate the levels hemoglobin and thus helps to prevent the onset of anemia of inflammation.

Preparation Note

The product is water soluble at a minimum of 5 mg/mL.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library