Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553211

Sigma-Aldrich

Rapamycin

≥95% (HPLC), solution, mTOR inhibitor, Calbiochem

Synonym(s):

InSolution Rapamycin, mTOR Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H79NO13
Molecular Weight:
914.17
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus

Quality Level

Assay

≥95% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

Selectively inhibits the phosphorylation and activation of p70 S6 kinase (IC50 = 50 pM). Prevents the translational activation of IGF-II. Also prevents resting T-cells from entering the cell cycle, but does not directly arrest cell cycle progression. Shown to inhibit later signaling events, such as p110Rb phosphorylation, p34cdc2 kinase activation, and cyclin A synthesis. Exhibits strong binding to FK-506 binding proteins. Also reported to induce apoptosis in a murine B-cell line, to inhibit lymphokine-induced cell proliferation at the G1 phase, and to irreversibly arrest Saccharomyces cerevisiae cells in the G1 phase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Mammalian target of rapamycin (mTOR)
Product does not compete with ATP.
Reversible: no
Target IC50: 50 pM inhibiting the mammalian target of rapamycin (mTOR) and blocking the subsequent activation of p70 S6 kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

2.5 mg/mL solution in DMSO (2.74 mM)

Reconstitution

Following initial use, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel M Wise et al.
Cells, 9(10) (2020-10-04)
Human adipose-derived stem cells (ASCs) show immense promise for treating inflammatory diseases, attributed primarily to their potent paracrine signaling. Previous investigations demonstrated that short-term Rapamycin preconditioning of bone marrow-derived stem cells (BMSCs) elevated secretion of prostaglandin E2, a pleiotropic molecule
Autophagy regulates sex steroid hormone synthesis through lysosomal degradation of lipid droplets in human ovary and testis.
Esmaeilian, et al.
Cell Death & Disease, 14, 342-342 (2023)
Tara Alpert et al.
Cell reports, 33(4), 108324-108324 (2020-10-29)
Nascent RNA sequencing has revealed that pre-mRNA splicing can occur shortly after introns emerge from RNA polymerase II (RNA Pol II). Differences in co-transcriptional splicing profiles suggest regulation by cis- and/or trans-acting factors. Here, we use single-molecule intron tracking (SMIT)
Clarissa Braun et al.
Cell stress, 5(12), 176-182 (2021-12-18)
Programmed cell death protein 4 (PDCD4) exerts critical functions as tumor suppressor and in immune cells to regulate inflammatory processes. The phosphoinositide 3-kinase (PI3K) promotes degradation of PDCD4 via mammalian target of rapamycin complex 1 (mTORC1). However, additional pathways that
Opeyemi S Adeniji et al.
Methods in molecular biology (Clifton, N.J.), 2442, 463-474 (2022-03-24)
The β-galactoside-binding protein Galectin-9 (Gal-9) functions as a double-edged sword during HIV infection. On the one hand, Gal-9 can reactivate HIV latently infected cells, the main barrier to achieving HIV eradication, making them visible to immune clearance. On the other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service