Skip to Content
MilliporeSigma
All Photos(1)

Documents

361550

Sigma-Aldrich

BIO

≥97% (HPLC), solid, GSK-3GSK-3α/β inhibitor, Calbiochem

Synonym(s):

GSK-3 Inhibitor IX, BIO, (2ʹ Z,3ʹ E)-6-Bromoindirubin-3ʹ-oxime, BIO, (2ʹZ,3ʹE)-6-Bromoindirubin-3ʹ-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10BrN3O2
CAS Number:
Molecular Weight:
356.17
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

GSK-3 Inhibitor IX, BIO, CAS 667463-62-9, is a cell-permeable, highly potent, selective, reversible, and ATP-competitive inhibitor of GSK-3α/β (IC₅₀ = 5 nM). Maintains self-renewal in human & mouse embryonic stem cells.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

purple

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

2-8°C

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miki Tokuoka et al.
PLoS genetics, 19(9), e1010953-e1010953 (2023-09-27)
How gene regulatory networks (GRNs) encode gene expression dynamics and how GRNs evolve are not well understood, although these problems have been studied extensively. We created a digital twin that accurately reproduces expression dynamics of 13 genes that initiate expression
Yanfei Deng et al.
Cellular reprogramming, 22(4), 217-225 (2020-07-17)
Wnt/β-Catenin signaling pathway plays an important role in maintaining self-renewal and pluripotency of human and mouse embryonic stem cells (ESCs). Activation of Wnt/β-Catenin signaling pathway by glycogen synthase kinase-3 (GSK3) inhibitor, the Wnt signaling agonist, could maintain the pluripotency of
Matthew Harder et al.
Developmental biology, 448(2), 136-146 (2018-10-06)
The gene regulatory networks underlying Ciona notochord fate specification and differentiation have been extensively investigated, but the regulatory basis for regionalized expression within the notochord is not understood. Here we identify three notochord-expressed genes, C11.331, C12.115 and C8.891, with strongly
Zhongde Ye et al.
NPJ aging and mechanisms of disease, 7(1), 4-4 (2021-02-10)
MicroRNAs play an important role in the regulation of T cell development, activation, and differentiation. One of the most abundant microRNAs in lymphocytes is miR-181a, which controls T cell receptor (TCR) activation thresholds in thymic selection as well as in
Masashi Hada et al.
Genes & development, 36(1-2), 84-102 (2022-01-08)
The placenta is a highly evolved, specialized organ in mammals. It differs from other organs in that it functions only for fetal maintenance during gestation. Therefore, there must be intrinsic mechanisms that guarantee its unique functions. To address this question

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service