Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00460

Sigma-Aldrich

Soyasapogenol B

≥98% (HPLC)

Synonym(s):

(3β,4β,22β)-Olean-12-ene-3,22,23-triol, Soyasapogenin B, Soyasapogenol I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O3
CAS Number:
Molecular Weight:
458.72
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1(C)C[C@@H](O)[C@]2(C)CC[C@@]3(C)[C@]4(C)CC[C@@]5([H])[C@](CO)(C)[C@@H](O)CC[C@]5(C)[C@@]4([H])CC=C3[C@]2([H])C1

InChI

1S/C30H50O3/c1-25(2)16-20-19-8-9-22-27(4)12-11-23(32)28(5,18-31)21(27)10-13-30(22,7)29(19,6)15-14-26(20,3)24(33)17-25/h8,20-24,31-33H,9-18H2,1-7H3/t20-,21+,22+,23-,24+,26+,27-,28+,29+,30+/m0/s1

InChI key

YOQAQNKGFOLRGT-UXXABWCISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Soyasapogenol B is a constituent of soya bean saponin that belongs to the family of oleanane triterpenes. It is known to have biofunctions in cell signaling, membrane integrity and stability, and fuel and energy storage. Soyasapogenol B may demonstrate hypocholesterolemic effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luping Wang et al.
Naunyn-Schmiedeberg's archives of pharmacology, 392(5), 551-563 (2019-01-05)
Clear cell renal cell carcinoma (ccRCC) is the most common type of human malignancies of the urological system. Soyasapogenol B (Soy B), an ingredient of soybean, has been found to exert anti-proliferative activities in vitro in human breast cancer cells.
Yan Zhang et al.
Pharmacological reports : PR, 72(5), 1370-1382 (2020-04-18)
This research aims to explore the effect of alpinumisoflavone (AIF) as an anti-cancer drug for the treatment of patients with hepatocellular carcinoma (HCC). Cell counting kit-8 (CCK-8) and colony formation assay were used to evaluate the viability of the cells
Lili Zhi et al.
Anatomical record (Hoboken, N.J. : 2007), 303(7), 1851-1858 (2019-10-04)
Soyasapogenol B (Soy B), a constituent of soybean, has been shown to exhibit antitumor activities against different types of cancers. However, to our knowledge, no studies so far have investigated the effect of Soy B in human laryngeal carcinoma. This
Madalina Neacsu et al.
Foods (Basel, Switzerland), 9(4) (2020-04-09)
Legumes are a source of health-promoting macro- and micronutrients, but also contain numerous phytochemicals with useful biological activities, an example of which are saponins. Epidemiological studies suggest that saponins may play a role in protection from cancer and benefit human
Teresa Herrera et al.
Journal of the science of food and agriculture, 99(6), 3157-3167 (2018-12-12)
Typical hydrolysis times of saponins generally do not take into consideration the effect of time on the degradation of the target compounds, namely sapogenins. When producing natural extracts, it should be borne in mind that conducting hydrolysis to yield a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service