Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SMB00434

Sigma-Aldrich

Jujuboside A

≥97% (HPLC)

Synonym(s):

(3β,16β,23R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→2)-O0[O-β-D-glucopyranosyl-(1→6)-O-[β-D-xylopyranosyl-(1→2)-β-D-glucopyranosyl-(1→3)]-α-L-arabinopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H94O26
CAS Number:
Molecular Weight:
1207.35
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Semen Ziziphus spinosa)

Quality Level

Assay

≥97% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC(C)=C[C@@H](C[C@@]1(O)C)O[C@]2(OC3)[C@@]1([H])[C@@]4([H])CC[C@@]5([H])[C@@]([C@]43C2)(C)CC[C@@]6([H])C(C)(C)[C@@H](O[C@@](OC[C@@H]7O)([H])[C@H](O[C@]8([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]8O)[C@H]7O[C@@](O9)([H])[C@H](O[C@@]%10([H])[C@H](O)[C@@H](O)[C@H]

InChI

1S/C58H94O26/c1-23(2)15-25-16-56(8,72)47-26-9-10-32-54(6)13-12-33(53(4,5)31(54)11-14-55(32,7)57(26)21-58(47,84-25)76-22-57)80-51-46(83-50-43(71)38(66)34(62)24(3)77-50)44(28(61)19-74-51)81-52-45(82-49-41(69)35(63)27(60)18-73-49)40(68)37(65)30(79-52)20-75-48-42(70)39(67)36(64)29(17-59)78-48/h15,24-52,59-72H,9-14,16-22H2,1-8H3/t24-,25-,26+,27+,28-,29+,30+,31-,32+,33-,34-,35-,36+,37+,38+,39-,40-,41+,42+,43+,44-,45+,46+,47-,48+,49-,50-,51-,52-,54-,55+,56-,57-,58-/m0/s1

InChI key

KVKRFLVYJLIZFD-OQVAHBRNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Jujuboside A (JuA) is a saponin isolated from Semen Ziziphus spinosa (SZS) used in Traditional Chinese Medicine.

Biochem/physiol Actions

Jujuboside A has been shown to be a major hypnotic-sedative and is used for treating insomnia and anxiety.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service