Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S2577

Sigma-Aldrich

Streptavidin High Capacity Coated Plates

8 well strip, clear

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

Quality Level

storage temp.

2-8°C

General description

These streptavidin-coated high capacity plates, in 8-well strip format, use a proprietary, unique coating technology to give greater (up to ~5-fold) biotin-binding capacity than standard streptavidin-coated plates. The 8-well strip format helps to minimize waste. These clear streptavidin-coated high capacity plates, made from polystyrene, are suitable for colormetric assays, and can be used in immunoassays and ELISA. Captured proteins can also be eluted for post-capture analysis by such various methods as MALDI, SDS-PAGE, or tagging technologies (e.g. ICAT, iTRAQ, mTRAQ).

Other Notes

96-well strip plate, with 8 wells per strip.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service