Skip to Content
MilliporeSigma

189730

Streptavidin

Synonym(s):

Streptavidin, Avidin, Streptomyces avidinii

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41116133
NACRES:
NA.41
MDL number:
Form:
lyophilized solid
Biological source:
(Streptomyces avidinii)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Streptomyces avidinii)

Quality Segment

form

lyophilized solid

specific activity

≥10 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

shipped in

ambient

storage temp.

−20°C

General description

A high affinity biotin binding protein (Kd approximately 10-15) that is stable over wide pH and temperature ranges. Each molecule of streptavidin binds 4 molecules of biotin. Streptavidin lacks the carbohydrate side chains present on avidin and has an isoelectric point nearer to neutrality (pI of 5.5-6.5 versus 10 for avidin). Consequently, streptavidin frequently exhibits lower levels of non-specific binding than avidin. The extinction coefficient of a 1% solution at 280 nm is 32.
A high-affinity biotin binding protein.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short-term storage. Aqueous stock solutions are stable for 6 months at 4°C and for up to 2 years at -20°C.
Reconstitute in H₂O or 1 mM NaHCO₃, pH 9.0 at twice the final concentration and then dilute with an equal volume of 2X buffer.

Other Notes

Eberle, J. and Seibl, R. 1992. J. Virol. Methods 40, 347.
Gudibande, S.R., et al. 1992. Mol. Cell. Probes. 6, 495.
Nesbitt, S.A. and Horton, M.A. 1992. Anal. Biochem. 206, 267.
Roth, J., et al. 1992. Histochemistry 98, 229.
Sano, T., et al. 1992. Science 258, 120.
Scouten, W.H. and Konecny, P. 1992. Anal. Biochem. 205, 313.
Srivastava, P., et al. 1992. Cytometry 13, 711.
One unit is defined as the amount of streptavidin required to bind 1.0 µg of D-Biotin at pH 7.0.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library