Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

STREPMAG-RO

Roche

Streptavidin Magnetic Particles

suspension

Synonym(s):

magnetic particles, streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

suspension

packaging

pkg of 10 mL (11641786001)
pkg of 2 mL (11641778001)

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

The Streptavidin Magnetic Particles are polydisperse paramagnetic particles that are designed for the fast separation of a variety of biotin-labeled molecules. The particles are supplied in a suspension that contains 10mg particles per ml in 50mM Hepes, 0.1% bovine serum albumin (BSA), 0.01% methyl-isothiazolone, pH 7.4.

Binding capacities per mg streptavidin magnetic particles:
>=1800 pmol free biotin
>150 pmol biotin-labeled oligonucleotide
>10 pmol biotin-labeled dsDNA fragment

Application

The Streptavidin Magnetic Particles are polydisperse paramagnetic particles that are designed for the fast separation of a variety of biotin-labeled molecules. The particles are supplied in a suspension that contains 10 mg particles per ml. The magnetic separation of biotin-labeled molecules has been successfully employed in a wide range of applications.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Schatte et al.
Bioconjugate chemistry, 27(6), 1484-1492 (2016-05-18)
Enzymes able to ligate biomolecules are emerging tools to generate site-specific bioconjugates. In this study we present a detection and screening method for bioconjugating enzymes which overcomes limitations of analytical methods such as HPLC or MS. These techniques are experimentally
Debolina Ray et al.
Cancer biology & therapy, 13(10), 848-857 (2012-07-13)
Stress treatment generally causes the post-translational modification and accumulation of the p53 protein, although the role of these aspects has not been always understood in relation to this protein's tumor suppressor activity. We analyzed these attributes of p53 in eight
Matthew T Balmer et al.
Cancer biology & therapy, 15(8), 1000-1012 (2014-05-08)
The chemotherapeutic agents doxorubicin (dox) or 5-fluorouracil (5FU) are used to treat cancer cells as they cause irreparable DNA damage, inducing these aberrant cells to undergo cell death. The mediator of this process is presumed to be in part the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service