Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK311

Sigma-Aldrich

Peroxide Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

Hydrogen Peroxide Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

cardiovascular diseases; aging/geriatric diseases; neonatal diseases; pediatric diseases; orthopedic diseases; neurological disorders; pulmonary disorders

storage temp.

2-8°C

General description

Peroxide (e.g. hydrogen peroxide H2O2), a reactive oxygen species produced through the metabolism of molecular oxygen, serves as both an intracellular signaling messenger and a source of oxidative stress. Hydrogen peroxide is generated in cells via multiple mechanisms such as the NOX-mediated ROS production by neutrophils and macrophages (respiratory burst) or by the dismutase of superoxide anions produced as a result of electron leak during mitochondrial respiration. Abnormal hydrogen peroxide production contributes to oxidative cell damage and the progression of diseases such as asthma, atherosclerosis, osteoporosis, and neurodegeneration.

Application

Peroxide Assay Kit has been used to measure hydrogen peroxide level in rat tissue homogenates.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the quantitative determination of peroxide concentration without any pretreatment in a variety of biological samples, such as serum, citrate-plasma, urine, cell lysates, and culture medium.

Principle

The Peroxide Assay Kit provides a simple and high-throughput adaptable assay for quantitative determination of peroxide concentration without any pretreatment in a variety of biological samples, such as serum, citrate-plasma, urine, cell lysates, and culture medium. This assay utilizes the chromogenic Fe3+ −xylenol orange reaction, in which a purple complex is formed when Fe2+ is oxidized to Fe3+ by peroxides present in the sample, generating a colorimetric (585 nm) result, proportional to the level of peroxide present. The optimized formulation reduces interference by substances in the raw samples. A concentration of 1 μM H2O2 equals 34 ng/mL or 34 ppb. The kit has a detection range of 0.2–30 μM (7–1,020 ng/mL) H2O2 in a 96 well format.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhiman Chakravarty et al.
Bioresource technology, 293, 122013-122013 (2019-09-09)
Here, we show the utility of a cyanobacterial biomass for overproduction and easy downstream processing of the thermostable protein KatB (a Mn-catalase). The nitrogen-fixing blue-green alga, Anabaena, was bioengineered to overexpress the KatB protein (An-KatB). Interestingly, pure An-KatB could be
Steffen Pabel et al.
Circulation research, 130(7), 994-1010 (2022-02-24)
Atrial fibrillation (AF) and heart failure often coexist, but their interaction is poorly understood. Clinical data indicate that the arrhythmic component of AF may contribute to left ventricular (LV) dysfunction. This study investigates the effects and molecular mechanisms of AF
Role of Cystathionine Gamma Lyase in Diabetic Nephropathy.
Anter A F, et al.
Journal of Advanced Biomedical and Pharmaceutical Sciences, 1(1 ), 33-40 (2018)
T O Ajiboye et al.
International journal of antimicrobial agents, 53(4), 467-473 (2018-12-12)
Zinc is a highly coveted redox-inactive micronutrient required for the growth and virulence of Acinetobacter baumannii. In this study, the role of the zinc uptake regulator Zur in the susceptibility and oxidative stress response of A. baumannii to antibiotics was
Han Xie et al.
Redox biology, 53, 102340-102340 (2022-05-25)
Generation of reactive oxygen species (ROS) are possibly induced by the crosstalk between mitochondria and endoplasmic reticula, which is physiologically important in apoptosis. Cytochrome c (Cyt c) is believed to play a crucial role in such signaling pathway by interrupting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service