Skip to Content
MilliporeSigma
All Photos(5)

Documents

MAK568

Sigma-Aldrich

Lipid Peroxidation (MDA) Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

MDA Assay, TBARS Assay, Thiobarbituric Acid Reactive Substance Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104

usage

sufficient for 100 colorimetric or fluorometric tests

Quality Level

detection method

colorimetric
fluorometric

relevant disease(s)

diabetes; cancer; cardiovascular diseases; neurological disorders; endocrinological disorders

storage temp.

−20°C

General description

Lipid peroxidation is the breakdown of lipids caused by oxidative damage, serving as a valuable marker for oxidative stress. Polyunsaturated lipids are particularly vulnerable to oxidative attack, often initiated by reactive oxygen species, triggering a well-defined chain reaction that yields key end products such as malondialdehyde (MDA). Uncontrolled lipid peroxidation has been implicated in the development of numerous diseases, including atherosclerosis, diabetes, and Alzheimer′s. The Lipid Peroxidation (MDA) Assay Kit is designed for measuring malondialdehyde (MDA) in a variety of samples including tissue, cells and plasma. Lipid peroxidation is determined by the reaction of MDA with thiobarbituric acid (TBA) to form a colorimetric (532 nm)/fluorometric (λex = 532/λem = 553 nm) product, proportional to the MDA present.

Features and Benefits

Highly sensitive, simple and high-throughput assay for measuring malondialdehyde (MDA) concentration. Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.

Click here to download a calculator excel file

Suitability

Suitable for the detection of malondialdehyde (MDA) in various biological samples such as cell and tissue culture, and plasma samples.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service