Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.10011

Supelco

Peroxide Test Strips, colorimetric

greener alternative

0.5-25 mg/L (H₂O₂), MQuant®, for use with MQuant® StripScan App

Synonym(s):

Peroxide chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Peroxide Test, colorimetric, 0.5-25 mg/L (H2O2), for use with MQuant® StripScan App, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: enzymatic reaction
reagent type: oxidant

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

0.5-25 mg/L (H2O2)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 0.5 - 2 - 5 - 10 - 25 mg/L (H2O2), MQuant®

Peroxide is an all purpose disinfectant which, due to its excellent antiviral, bactericidal and fungicidal action, is widely used in the food industry. Here, the MQuant® test strips are ideal for controlling the residual peroxide after disinfection. Peroxidase transfers peroxide oxygen to an organic redox indicator. This produces a blue oxidation product. The peroxide concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the peroxide concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Peroxide test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Application of copper (I) selective ligands for PET imaging of reactive oxygen species: This research uses peroxide detection in biomedical imaging to trace reactive oxygen species in vivo, aiding in the diagnosis and study of diseases associated with oxidative stress (Tada et al., 2024).


  • Optimization of Pressurized Liquid Extraction (PLE) Parameters for Extraction of Bioactive Compounds: This study employs peroxide tests in the extraction of bioactive compounds from plant materials, enhancing the efficiency of natural product extraction processes (Chatzimitakos et al., 2024).


  • Highly Sensitive, Easy-to-Use, One-Step Detection of Peroxide-, Nitrate- and Chlorate-Based Explosives: Developing highly sensitive detection methods for explosives using electron-rich Ni porphyrins, this application serves crucial roles in safety and security fields, demonstrating the versatility of peroxide tests (Brockmann et al., 2024).


Linkage

Replaces: 10011-1; 10011

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Protocols

Preparation of a standard solution for Hydrogen peroxide

Related Content

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service