Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z683299

XPell indicating pellets for peroxides prevention

CE for organic solvents

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

packaging

pack of 80 g

manufacturer/tradename

XploSafe 3002

Looking for similar products? Visit Product Comparison Guide

General description

XPell CE indicating pellets for peroxides prevention in organic solvents. Allows proper management of peroxide forming solvents and eliminates the possibility of serious accidents resulting from peroxide buildup.

Directions: For each liter of solvent, add one tablespoon (approx 10 g) of XPell CE. XPell CE neutralizes peroxides present in the solvent and peroxides that form over time. The solvent is protected as long as Xpell CE pellets remain greenish gray. A color change to a yellow/pale color indicates that the protection is exhausted. Once this change occurs, add more pellets to the solvent until the color of the additional pellets remain dark greenish gray.

Note: XPell CE works by converting reactive, explosive peroxides into nominally non-reactive alcohols that are not likely to interfere with UV-VIS measurements, most analytical methods, and organic reactions and separations. For most solvents, the purity will remain high (impurities less than 0.1%) for over 12 months even with continuous exposure to oxygen and light. For many analytical uses, an open bottle with XPell CE is expected to remain viable (impurities 100 ppm or less) for over two months or longer depending on the storage conditions. Given the non-reactive nature of the byproducts, many applications will be able to use the solvent protected with XPell CE for a much longer period of time. The user is encouraged to verify that the products generated from peroxide neutralization do not cause any interference with their chosen analytical method.

80 grams (sufficient for treating up to 8 L of solvent)

Legal Information

XPell is a trademark of XploSafe, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service