Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1919

Sigma-Aldrich

Monoclonal Anti-Macrophages antibody produced in mouse

clone LN-5, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LN-5, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400 using formalin-fixed, paraffin-embedded human tonsil sections

isotype

IgM

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-Human Macrophages (mouse IgM isotype) is derived from the LN-5 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Macrophages are widely distributed throughout the body. They belong to the mononuclear phagocytes family. Macrophages have abundant cytoplasm and are also termed histiocyte.

Immunogen

deparaffinized human peripheral blood mononuclear cells

Biochem/physiol Actions

Macrophages play a crucial role in body defense mechanisms and have phagocytic functionality. They are heterogeneous in morphology, cytochemistry, function and surface antigens. Monoclonal antibodies to macrophage and histiocyte surface or cytoplasmic antigens are important for the immunodiagnosis of both benign and malignant histiocytic lesions. They are useful in the investigation of macrophage differentiation and maturation and the diagnosis of various inflammation.

Physical form

The product is provided as ascites fluid with 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Slava Epelman et al.
Immunity, 41(1), 21-35 (2014-07-19)
Macrophages are distributed in tissues throughout the body and contribute to both homeostasis and disease. Recently, it has become evident that most adult tissue macrophages originate during embryonic development and not from circulating monocytes. Each tissue has its own composition
Caoimhe Egan et al.
Seminars in diagnostic pathology, 35(1), 20-33 (2017-11-19)
Benign and malignant proliferations of histiocytes and dendritic cells may be encountered in lymph nodes. Reactive histiocytic and dendritic cell infiltrates occur in response to diverse stimuli and in addition to causing lymphadenopathy, may be present unexpectedly in lymph nodes
Daisuke Hirayama et al.
International journal of molecular sciences, 19(1) (2017-12-30)
Macrophages are effector cells of the innate immune system that phagocytose bacteria and secrete both pro-inflammatory and antimicrobial mediators. In addition, macrophages play an important role in eliminating diseased and damaged cells through their programmed cell death. Generally, macrophages ingest
Siamon Gordon et al.
BMC biology, 15(1), 53-53 (2017-07-01)
Macrophages are present in all vertebrate tissues, from mid-gestation throughout life, constituting a widely dispersed organ system. They promote homeostasis by responding to internal and external changes within the body, not only as phagocytes in defence against microbes and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service