Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3215

Sigma-Aldrich

JE/MCP-1 (CCL2) from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCL2, JE, MCAF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

13.8 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CCL2(20296)

General description

The monocyte chemoattractant proteins (MCPs) belong to the C-C chemokine family, and signal through C-C motif chemokine receptor-2 (CCR2) and, with the exception of MCP-1 or chemokine (C-C motif) ligand 2 (CCL2) , other CCR receptors. The MCP family cross reacts across species. The gene encoding CCL2 is localized on mouse chromosome 11. Recombinant murine JE is a 13.8kDa protein containing 125 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.

Biochem/physiol Actions

The monocyte chemoattractant proteins (MCPs) chemo attract and activate monocytes, activated T cells, basophils, natural killer (NK) cells and immature dendritic cells. It has a role in the survival and proliferation of microglia. Chemokine (C-C motif) ligand 2 (CCL2) has been shown to activate GTPases like cell division control protein 42 (CDC42) and Ras-related C3 botulinum toxin substrate 1 (Rac1).

Sequence

QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for a cluster of genes on chromosome 17q11-q21 controlling susceptibility to tuberculosis and leprosy in Brazilians.
Jamieson SE
Genes and Immunity, 5(1), 46-57 (2004)
Vascular Stem/Progenitor Cell Migration Induced by Smooth Muscle Cell-Derived Chemokine (C-C Motif) Ligand 2 and Chemokine (C-X-C motif) Ligand 1 Contributes to Neointima Formation.
Yu B
Stem Cells, 34(9), 2368-2380 (2016)
Rollins, R.J., et al.
Cytokine Reference, 1145-1160 (2001)
Magdaléna Vaváková et al.
Scientific reports, 13(1), 10559-10559 (2023-06-30)
Salt-inducible kinase 2 (SIK2) is highly expressed in white adipocytes, but downregulated in individuals with obesity and insulin resistance. These conditions are often associated with a low-grade inflammation in adipose tissue. We and others have previously shown that SIK2 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service