Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

375645

Sigma-Aldrich

HePTP Inhibitor, ML119

The HePTP Inhibitor, ML119 controls the biological activity of HePTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

HePTP Inhibitor, ML119, (Z)-2-(5-(7-Bromo-3-oxo[1,3]thiazolo(2ʹ,3ʹ:2,3)imidazo[4,5-b]pyridin-2(3H)-ylidene)methyl)furan-2-yl)benzoic acid, PTPN7 Inhibitor, ML119, CID1357397, CD45 Inhibitor IV, Dusp6/MKP3 Inhibitor II, EC-PTP/PCPTP1 Inhibitor II, Lyp Inhibitor II, PTP Inhibitor XXX, PTP-SL Inhibitor II, PTP1B Inhibitor VIII, PTPN2/TCPTP Inhibitor III, SHP1 Inhibitor VIII, SHP2 Inhibitor V, STEP Inhibitor, VHR Inhibitor III, CD45 Inhibitor IV, Dusp6/MKP3 Inhibitor II, EC-PTP/PCPTP1 Inhibitor II, Lyp Inhibitor II, PTP Inhibitor XXX, PTP-SL Inhibitor II, PTP1B Inhibitor VIII, PTPN2/TCPTP Inhibitor III, SHP1 Inhibitor VIII, SHP2 Inhibitor V, STEP Inhibitor, VHR Inhibitor III, (<

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10BrN3O4S
CAS Number:
Molecular Weight:
468.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

210 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

green-yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

BrC1=CC2=C(N(C(/C(S3)=C/C4=CC=C(C5=CC=CC=C5C(O)=O)O4)=O)C3=N2)N=C1

General description

A cell-permeable oxothiazolo-imidazo-pyridinylidene-benzoate that competitively blocks the activity of hematopoietic protein tyrosine phosphatase (HePTP) (IC50 = 210 nM; Ki = 211 nM) and thereby induces sustained hyperactivation of ERK1/2 and p38 MAP kinase (in OKT3-stimulated Jurkat TAg T cells at 4 µM, and in mouse leukocytes (3 mg/kg, i.p.)) without affecting MEK and Akt. Shown to interact with the catalytic pocket of HePTP loop and act as a pTyr mimetic. Displays adequate selectivity over HePTP-H237A, MKP-3, VHR, STEP, PTP-SL, PTP1B, LYP, TCPTP, SHP1, SHP2, CD45 and PAR phosphatases (IC50 = 2.8, 5.2, 1.9, 6.3, 1.1, 1.7, 1.9, 1.0, 2.5, 1.1, 3.1 and 73 µM, respectively). Exhibits greater microsomal stability (t1/2 = 50 min in rat liver microsomes).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HePTP/PTPN7
Reversible: yes
Secondary Target
other phosphatases
Target Ki: 211 nM for HePTP

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sergienko, E., et al. 2012. ACS Chem. Biol.7, 367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service