Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M047000

Sigma-Aldrich

Cyanine 3 Phosphoramidite

Synonym(s):

2-[3-[1,3-Dihydro-1-[3-(4-methoxytriphenylmethoxy)propyl]-3,3-dimethyl-2H-indol-2-ylidene]-1-propen-1-yl]-1-[3-[N,N-diisopropylamino(2-cyanoethoxy)phosphinoxy]propyl]-3,3-dimethyl-3H-indolium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H70N4O4P
CAS Number:
Molecular Weight:
918.17
UNSPSC Code:
41116105

product line

Proligo Reagents

Quality Level

Assay

≥90% (31P-NMR)
≥95.0% (@549nm, reversed phase HPLC)

color

dark red, 0.1M in Acetonitrile

turbidity

clear

λ

conforms (UV/VIS Identity)

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service