Skip to Content
MilliporeSigma
All Photos(1)

Documents

LVCAS9BST

Sigma-Aldrich

CAS9 Blasticidin Lenti Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

packaging

vial of 200 μL

Quality Level

concentration

≥1 x 10 6 VP/ml virus (via p24 assay)

selection

blasticidin

shipped in

dry ice

storage temp.

−70°C

General description

Ready-to-use Cas9 lentiviral particles are ideal for transducing a wide range of cell lines. The Cas9 Blasticidin Lentiviral particles efficiently integrate Cas9 into the genomes of both dividing and non-dividing cell lines for use with CRISPR genome editing. Stably integrate Cas9 and Blast using the convenient, ready-to-use lentiviral particles while avoiding the tedious lentivirus production process.

Application

Functional Genomics/Screening /Target Validation/Genome Editing

Components

200 ul of purified lentiviral particles (minimum of 1 X 106 TU/ml via a p24 assay) expressing Cas9 only with blasticidin resistance.

Linkage

Click here to order gRNA plasmid and virus.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service