Skip to Content
MilliporeSigma
All Photos(1)

Documents

CAS9BST

Sigma-Aldrich

CAS9 Blasticidin Lenti Plasmid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

recombinant

expressed in E. coli

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

selection

ampicillin
blasticidin

shipped in

dry ice

storage temp.

−20°C

General description

This product is a blasticidin resistant lenti-Cas9 plasmid for generation of lentiviral particles. Use Sigma′s lentiviral Cas9 reagents to efficiently generate stable cell lines expressing Cas9 for CRISPR genome editing. Sigma′s lenti-Cas9 plasmid is one part of a two-vector CRISPR system with individual Cas9 and gRNA expression vectors. Please see the link below to order gRNA.

Application

Functional Genomics/Screening /Target Validation/Genome Editing

Components

1ug of plasmid DNA, EF1a-Cas9-2A-Blasticidin Lenti

Linkage

Click here to order gRNA plasmid and virus.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service