Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HUMANCD4-0002099

Sigma-Aldrich

Frozen Human Normal CD4+ Helper T Cells

human peripheral blood (CD4+ Helper T Cells)

Synonym(s):

CD4+ Cells, Helper T Cells, Immune Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Frozen Human Normal CD4+ Helper T Cells, 10 Million Cells/per vial

biological source

human peripheral blood (CD4+ Helper T Cells)

form

frozen liquid

packaging

vial of 10 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products from BioIVT.

T cells are lymphocytes, a subset of of white blood cells, that are important to adaptive immunity. T cells orginate in the bone marrow and mature in the thymus where they expand and differentiate into helper, regulatory, or cytotoxic T cells or become memory T cells. CD4+ T cells are a subset of T cells that make up the majority of T cells. Although there are distinct subsets of CD4+ cells that have various functions, CD4+ T cells are primarily considered helper T cells. CD4+ helper T cells are isolated from mononuclear cells via immunomagnetic separation. CD4+ Pan T cells can be isolated from a variety of tissue sources including peripheral blood and lymphoid tissues.

Used for a variety of applications, T cells are frequently utilized in drug discovery/development, assay validation/development, cell therapy tissue engineering, mixed lymphocyte reaction assays, and other immunology applications.

Packaging

10 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service