Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

685194

Sigma-Aldrich

2-Methyl-4-cyanophenylboronic acid

≥95%

Synonym(s):

(4-Cyano-2-methylphenyl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8BNO2
CAS Number:
Molecular Weight:
160.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

287-305 °C

SMILES string

Cc1cc(ccc1B(O)O)C#N

InChI

1S/C8H8BNO2/c1-6-4-7(5-10)2-3-8(6)9(11)12/h2-4,11-12H,1H3

InChI key

RGCVYEOTYJCNOS-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant involved in:
  • Reactions with (cyanomethylphenyl)boronic acid
  • Synthesis of oligo-boronic acid receptors with improved structural and electronic properties
  • Syntesis of aminoalkoxybiphenylnitriles for use as histamine-3 receptor ligands

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service