Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7909

Sigma-Aldrich

HU-210

solid (air sensitive)

Synonym(s):

(6aR-trans-3-(1, 1-Dimethylheptyl)-6a,7,10,10a-tetrahydro-1-hydroxy-6,6-dimethyl-6H-dibenzo[b,d]pyran-9-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H38O3
CAS Number:
Molecular Weight:
386.57
Beilstein:
4298162
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid (air sensitive)

drug control

USDEA Schedule I; stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

solubility

DMSO: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@@]12CC(CO)=CC[C@@]1([H])C(C)(C)Oc3cc(cc(O)c23)C(C)(C)CCCCCC

InChI

1S/C25H38O3/c1-6-7-8-9-12-24(2,3)18-14-21(27)23-19-13-17(16-26)10-11-20(19)25(4,5)28-22(23)15-18/h10,14-15,19-20,26-27H,6-9,11-13,16H2,1-5H3/t19-,20-/m1/s1

InChI key

SSQJFGMEZBFMNV-WOJBJXKFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cannabinoid receptor agonist.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Martín-Calderón et al.
European journal of pharmacology, 344(1), 77-86 (1998-05-15)
In the present study we have characterized the effects of the acute administration of the synthetic cannabinoid (-)-11-hydroxy-delta8-tetrahydrocannabinol-dimethylheptyl (HU-210, 4, 20 and 100 microg/kg), on the secretion of prolactin, growth hormone, luteinizing hormone, follicle-stimulating hormone, adrenocorticotropic hormone and corticosterone in
Bob Roozenbeek et al.
Critical care medicine, 40(5), 1609-1617 (2012-04-19)
The International Mission on Prognosis and Analysis of Clinical Trials and Corticoid Randomisation After Significant Head injury prognostic models predict outcome after traumatic brain injury but have not been compared in large datasets. The objective of this is study is
Valentina Chiodi et al.
Neurobiology of disease, 45(3), 983-991 (2011-12-31)
Cannabinoid CB1 receptors (CB1Rs) are known to be downregulated in patients and in animal models of Huntington's disease (HD). However, the functional meaning of this reduction, if any, is still unclear. Here, the effects of the cannabinoid receptor agonist WIN
Unyong Kim et al.
Journal of pharmaceutical and biomedical analysis, 64-65, 26-34 (2012-03-13)
(6aR,10aR)-9-(Hydroxymethyl)-6,6-dimethyl-3-(2-methyloctan-2-yl)-6a,7,10,10a-tetrahydrobenzo[c]chromen-1-ol (HU-210) is a synthetic cannabinoid, with a classical cannabinoid structure similar to Δ(9)-tetrahydrocannabinol (Δ(9)-THC). In this study, the in vitro metabolism of HU-210 was investigated in human liver microsomes to characterize associated phase I metabolites. HU-210 was incubated with
Lidia Martín-Couce et al.
Angewandte Chemie (International ed. in English), 51(28), 6896-6899 (2012-06-13)
Receptors made visible: The described biotin-tagged small-molecule probes with excellent affinities for the CB(1) and CB(2) cannabinoid receptors (CB(1)R and CB(2)R) enable direct visualization of these receptors in native cellular systems, including neurons, microglia, and immune cells. This method could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service