Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H17D6ClN6O2
Molecular Weight:
456.96
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
pharmaceutical (small molecule)
format
single component solution
storage temp.
−20°C
General description
Suvorexant, sold under the trade name Belsomra, is a medication for the treatment of insomnia, approved for sale by the U.S. Food and Drug Administration (FDA) in 2014. The U.S. Drug Enforcement Administration (DEA) has scheduled this substance as controlled substance, as it may lead to physical or psychological dependence. This stable-labeled internal standard is suitable for use in quantitation of suvorexant levels by GC/MS or LC/MS for applications in clinical toxicology, therapeutic drug monitoring or urine drug testing.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
49.5 °F - closed cup
Flash Point (°C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


