Skip to Content
MilliporeSigma

D8174

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F

≥98% (HPLC), solution, thromboxane A2 agonist

Synonym(s):

U-46619, U46619

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Beilstein/REAXYS Number:
4267334
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate

form

solution

Quality Segment

concentration

10 mg/mL in methyl acetate

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\[C@H]1C2CC(CO2)[C@@H]1C\C=C/CCCC(O)=O

InChI

1S/C21H34O4/c1-2-3-6-9-17(22)12-13-19-18(16-14-20(19)25-15-16)10-7-4-5-8-11-21(23)24/h4,7,12-13,16-20,22H,2-3,5-6,8-11,14-15H2,1H3,(H,23,24)/b7-4-,13-12+/t16?,17-,18-,19+,20?/m0/s1

InChI key

LQANGKSBLPMBTJ-REGKDVDGSA-N

General description

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F2α is an endoperoxide/thromboxane analog.

Application

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F2α has been used:
  • to induce aortic smooth muscle (SM) contraction in mice deficient in myosin light chain 9 (Myl9) gene
  • to induce contraction as part of vascular reactivity experiments using mice aorta
  • as a thromboxane/prostaglandin agonist to study the effect of dithiothreitol (DTT) on mice arterial vessel viability

Biochem/physiol Actions

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F2α is a thromboxane A2 agonist. It is useful in in vitro experiments to induce shape change, aggregation, secretion, phosphoinositide hydrolysis, and protein phosphorylation in platelets. It also causes an elevation in the cytosolic Ca++ levels slightly affecting the cyclic adenosine monophosphate (cAMP) formation. It acts as a potent stimulator of vascular cell adhesion molecule-1 (VCAM-1), which mediates leukocyte adhesion to the endothelium.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

14.0 °F

flash_point_c

-10 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library