Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0480-01

Epoxy Activated Sepharose 6B

Cytiva 17-0480-01, pack of 15 g

Synonym(s):

Epoxy Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 15 g

manufacturer/tradename

Cytiva 17-0480-01

matrix

6% agarose

particle size

45-165 μm

average diameter

90 μm

ligand dependent

2-14

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Epoxy-activated Sepharose 6B is a pre-activated medium for immobilization of various ligands including sugars through coupling of hydroxy, amino or thiol groups on the ligand to Sepharose 6B via a 12-atom hydrophilic spacer arm.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service