Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC (p)
Citations:
4
biological source
rabbit
conjugate
unconjugated
antibody form
Ig fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (236A-14), vial of 0.5 mL concentrate (236A-15), bottle of 1.0 mL predilute (236A-17), vial of 1.0 mL concentrate (236A-16), bottle of 7.0 mL predilute (236A-18)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200-1:500
control
colon
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
Gene Information
human ... CEACAM5(1048)
General description
Anti-CEA is employed essentially as a tool to assist in the distinction between adenocarcinoma and epithelioid malignant mesotheliomas, along with other markers such as those against calretinin, CK 5&6, CD15, HBME-1, MOC-31, and Ber-EP4. Another suggested use of anti-CEA is to immunophenotype various metastatic adenocarcinomas as a means of identifying their origin within a panel of different markers. Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix. Polyclonal anti-CEA is useful in staining hepatocellular carcinoma in a canalicular pattern.
Physical form
Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide
Preparation Note
Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.
Other Notes
CEA Positive Control Slides, Product No. 236S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).
For Technical Service please contact: 800-665-7284 or email: [email protected]
Legal Information
Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P W Shield et al.
American journal of clinical pathology, 105(2), 157-162 (1996-02-01)
The contribution of immunocytochemical (ICC) staining to the diagnosis of a range of cytologic specimens reported over a 20-month period was retrospectively assessed. A total of 194 cases (1.6% of total workload) were stained for diagnostic purposes in this period.
A Alkushi et al.
Virchows Archiv : an international journal of pathology, 442(3), 271-277 (2003-03-21)
Adenocarcinomas of the uterine cervix show a wide range of morphological features, and can be confused with endometrial adenocarcinoma in biopsy or curetting specimens. The objective of this study was to use tissue microarray technology to evaluate the immunoprofile of
Carl Morrison et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 15(12), 1279-1287 (2002-12-14)
The distinction of hepatocellular carcinoma (HCC) from metastatic adenocarcinoma (MA) and cholangiocarcinoma (CC) in some cases requires the use of immunohistochemistry. CD10 has recently been suggested as a useful stain for HCC. We directly compared CD10 with other immunohistochemical markers
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 236A-16 | 0841683129080 |
| 236A-15 | 0841683129073 |
| 236A-14 | 0841683129066 |
| 236A-17 | 0841683129097 |
| 236A-18 | 0841683129103 |