Skip to Content
MilliporeSigma

6B100

Sepharose 6B

6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)

Synonym(s):

Agarose, Beaded

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
41106500
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (marine)

form

suspension

technique(s)

affinity chromatography: suitable

matrix

6% agarose

particle size

45-165 μm (wet)

pore size

10,000-1,000,000 Da fractionation range (dextrans), 10,000-4,000,000 Da fractionation range (globular proteins)

pH

4—9

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@@H](O3)O[C@H]4[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]5[C@@H]6CO[C@H]5[C@H](O)[C@H](O)O6)[C@@H]4O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

6B100-1L′s update product number is GE17-0110-01

Other Notes

Beaded agarose for fractionating molecules of high molecular weight. Cross-linked beaded agarose is more resistant to denaturing conditions, and thus offers more versatility in the choice of sample buffer and eluent. The approximate % agarose concentration is indicated by the product name (Ultrogel A%).

Legal Information

Sepharose is a trademark of Cytiva


Still not finding the right product?

Explore all of our products under Sepharose 6B


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 - 109.4 °F

flash_point_c

38 - 43 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library