Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1924

Sigma-Aldrich

α-1→(2,3,4)-Fucosidase solution from Xanthomonas sp.

buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

buffered aqueous solution

Quality Level

specific activity

≥0.5 units/mg protein

packaging

vial of 0.004 unit

shipped in

wet ice

storage temp.

2-8°C

Unit Definition

One unit will hydrolyze 1 μmole fucose from 3-fucosyllactose per min at pH 5.0 at 37 °C.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service