Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK444

Sigma-Aldrich

α-L-Fucosidase Assay Kit

Sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

input

tissue
serum
plasma
cell lysate

detection method

colorimetric

relevant disease(s)

cancer, diabetes; endocrinological disorders, hepatitis; gastrointestinal diseases

storage temp.

2-8°C

General description

The enzyme α-L-Fucosidase (AFU), encoded by the FUCA1 gene in humans, is responsible for breaking down L-Fucose. Genetic deficiency in this enzyme leads to fucosidosis, a neurovisceral storage disease characterized by fucose accumulation. Reduced serum activity of fucosidase is associated with ovarian carcinoma, while elevated activity is linked to conditions such as diabetes, hyperthyroidism, cirrhosis, and hepatitis. Additionally, increased AFU activity has been observed in various types of carcinomas including lung, breast, stomach, ovary, uterus, and liver cancers.

Application

The α-L-Fucosidase Assay Kit may be used for:
  • Fucosidosis Research
  • Cancer Research
  • Endocrinology Research

Features and Benefits

  • Efficient Process: Experience a simplified process with our kit, requiring only one working reagent and a 20-minute assay reaction, saving valuable time and effort in your workflow.
  • Expanded Detection Capability: Detect α-ʟ-Fucosidase levels precisely across a wide range, from 1 to 100 U/L, using just 10 μL of sample in a 96-well plate, allowing for comprehensive analysis with minimal sample volume.
  • Compatibility with High-Throughput Systems: Seamlessly integrate our kit into high-throughput handling systems, ensuring streamlined operations and precise results, enhancing efficiency in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of α-L-Fucosidase activity in plasma, serum, tissue, cell lysate, etc.

Principle

The non-radioactive, colorimetric α-L-Fucosidase Assay Kit is based on the cleavage of 4-nitrophenol from the synthetic substrate. Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at A405 nm after addition of the stop reagent is directly proportional to the enzyme activity.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service