Key Documents
EP0030124847
Eppendorf® cap strips for PCR tubes
flat, pkg of 120 ea, 10 x 12 strips
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
flat
feature
autoclavable 120 °C
flat
packaging
pkg of 120 ea
manufacturer/tradename
Eppendorf® 0030124847
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)