Key Documents
EP0030124901
Eppendorf® FAST PCR-Tube Strips
capacity 0.1 mL, pack of 120 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
cap (plug seal)
colorless tube
conical bottom
feature
PCR clean
graduations: no
lid: no
neck thread Push fit
8 tube-strips, separate cap strip
packaging
pack of 120 ea
manufacturer/tradename
Eppendorf® 0030124901
technique(s)
PCR: suitable
capacity
0.1 mL
General description
The Eppendorf Fast PCR Tube Strips made of Polyethylene transfer heat faster than the tubes made of traditional plastics. The Fast PCR Tube Strips are particularly suited to scientists using Fast PCR Reagent Kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates - such as the Eppendorf Mastercycler X50 family.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG