MilliporeSigma
V0756
Vortex adapter for microcentrifuge tubes
capacity, 20 tubes
Synonym(s):
370800020, H37080-0020, agitator adapter, stirrer adapter, stirrer adapter for tubes, vortex adapter
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121702
NACRES:
NB.73
eCl@ss:
32119210
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
capacity 20 tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H37080-0020
General description
For use with any vortex mixer equipped with a rubber pad or test tube cup. The device holds 1.5 mL tubes in a disk mounted on a shaft. A sliding plate holds down the tube caps when in use. Provides rapid and complete mixing of many samples at once.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)