Key Documents
CLS6795400D
Corning® hotplate with digital display
AC/DC input 120 V AC, US 3-pin plug, L × W 5 in. × 7 in.
About This Item
UNSPSC Code:
41102406
NACRES:
NB.76
material
ceramic plate
sterility
non-sterile
packaging
case of 1
manufacturer/tradename
Corning 6795-400D
availability
not available in EU
parameter
5-550 °C temp. range
L × W
5 in. × 7 in.
color
none
General description
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Microprocessor maintains consistent and repeatable temperature settings.
  • Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
  • Separate temperature sensor provides power cut off if unit overheats
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
  • 2 year warranty
Legal Information
Corning is a registered trademark of Corning, Inc.