Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS6795KIT

Corning® digital display hot plate and stirring hot plate accessory kit

accessory kit is designed for all Corning digital display hot plates and stirring hot plates, 1/cs

Synonym(s):

Corning Hot Plate Kit, Digital Hot Plate Kit, Stirring Hot Plate Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.73

sterility

non-sterile

packaging

case of 1
pack of 1

manufacturer/tradename

Corning 6795KIT

color

none

compatibility

all Corning digital display hot plates and stirring hot plates

General description

This accessory kit is designed for all Corning digital display hot plates and stirring hot plates (Models CLSPC400D, CLSPC420D, PC-600D, CLSPC620D). It is voltage independent and includes a temperature controller (CLS6795PR) that controls the temperature of liquids up to 200ºC, a two piece, 5/16 in. (0.8 cm) diameter, 18 in. (45.7cm) long support rod (CLS440129) and magnetic stir bar retriever (CLS6970SR).

  • Use with models: CLSPC400D, CLSPC420D, CLSPC600D, CLSPC620D
  • Includes a temperature controller (CLS6795PR), a two piece, 5/16 in. (0.8 cm) diameter, 18 in. (45.7cm) long support rod (CLS440129) and magnetic stir bar retriever (CLS6970SR). (Boss Head clamp (CLS440140) and holding rod (CLS440141) are pictured, but are NOT included in this kit)

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service