Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS6795620D

Corning® hotplate and stirrer with digital display

AC/DC input 120 V AC, US 3-pin plug, plate L × W 10 in. × 10 in.

Synonym(s):

magnetic stirrer, magnetic stirrers, stirrers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.72

sterility

non-sterile

packaging

case of 1

manufacturer/tradename

Corning 6795-620D

availability

not available in EU

parameter

5-550 °C temp. range
60-1150 rpm speed

plate L × W

10 in. × 10 in.

color

none

Looking for similar products? Visit Product Comparison Guide

General description

  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Microprocessor maintains consistent and repeatable temperature and stir speed settings.
  • Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
  • Separate temperature sensor provides power cut off if unit overheats
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service