Key Documents
CLS3788
Corning® 96 Well Clear Polystyrene Microplate
round bottom clear, polystyrene, bag of 20 ×, sterile, lid
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
polystyrene
round bottom clear
size:
96 wells
Pricing and availability is not currently available.
material
clear bottom
polystyrene
round bottom clear
sterility
sterile
feature
lid
skirt
packaging
case of 100
bag of 20 ×
manufacturer/tradename
Corning 3788
size
96 wells
well volume
330 μL
working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200 ng IgG/cm2
  • Total volume is 330 μL; recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
Note: Total well volume is 330 μL
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)