Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3879

Sigma-Aldrich

Corning® 96 well clear polypropylene corner notch microplate

round bottom clear, polypropylene, individually wrapped, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13
material:
clear bottom
polypropylene
round bottom clear
maximum volume:
320 μL
size:
96 wells
working volume:
200 μL
sterility:
sterile
binding type:
non-treated surface
feature:
individually wrapped
lid

material

clear bottom
polypropylene
round bottom clear

sterility

sterile

feature

individually wrapped
lid

packaging

case of 50

manufacturer/tradename

Corning 3879

maximum volume

320 μL

size

96 wells

working volume

200 μL

color

clear

suitability

suitable for (for cell culture applications; growing cells in stationary suspension)

binding type

non-treated surface

General description

96 well standard polypropylene plate designed for growing cells in stationary suspension or other cell culture applications where reduced cell attachment is desired.

  • Designed for growing cells in stationary suspension or other cell culture applications where reduced cell attachment is desired
  • Round bottom wells with 330 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Individually wrapped

Note: This is the suggested replacement for CLS3790

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S B Costa et al.
Diagnostic microbiology and infectious disease, 96(3), 114870-114870 (2020-01-21)
Continuous hemodialysis system monitoring is necessary to prevent microorganism growth and health problems. This study evaluates single- and dual-species biofilm formation in microtiter plates by using dialysis solutions under aerobiosis or 5% CO2 atmosphere. Escherichia coli, Pseudomonas aeruginosa, Staphylococcus epidermidis
Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service