Skip to Content
MilliporeSigma

CLS3799

Corning® 96 Well TC-Treated Microplates

size 96 wells, clear, polystyrene, round bottom, case of 50 (individually wrapped), sterile, lid

Synonym(s):

cell culture plate, tissue culture plates, well plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122107
NACRES:
NA.75
eCl@ss:
32190102
Material:
clear , clear bottom, polystyrene , round bottom
Size:
96 wells
Sterility:
sterile; γ-irradiated by SER-TAIN process, sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid, skirt
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , clear bottom, polystyrene , round bottom

sterility

sterile; γ-irradiated by SER-TAIN process, sterile

quality

tissue-culture treated

feature

lid, skirt

packaging

case of 50 (individually wrapped)

manufacturer/tradename

Corning 3799

size

96 wells

well diam.

6.4 mm

well volume

330 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Features and Benefits

  • Corning® 96 well microplates support various application requirements.
  • Tissue culture-treated (TC-treated) polystyrene surface is used for the attachment and growth of anchorage-dependent cells.
  • The round bottom is usually the best choice for washing, retrieving and sample auto-centering.

Legal Information

Corning is a registered trademark of Corning, Inc.
SER-TAIN is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library