Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3632

Corning® 96 Well White Polystyrene Microplate

flat bottom clear, white polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

flat bottom clear
round clear wells
white polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3632

size

96 wells

well volume

360 μL

working volume

75-200 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescent and enabling readings down to 340 nm
  • Designed to reduce well-to-well crosstalk during luminescent assays
  • Can be used for both top and bottom reading instruments
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200 ng IgG/cm2
  • Without lids, lids supplied separately
  • Total volume is 360 μL; recommended working volume of 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service